Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile, 75-05-8, MFCD00001878, 1X4L
Catalog No. 50156220EA
Encompass
Linear Formula: CH3CN, Molecular Weight: 41.05, for HPLC, gradient grade, ≥99.9%, Synonym: ACN, Methyl cyanide. Acetonitrile (MeCN) may be used as an extraction solvent during the quantification of pesticide residues in fruits and vegetables.
Catalog No. 50156220EA Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348514L
Linear Formula: CH3CN, Molecular Weight: 41.05, for HPLC, gradient grade, ≥99.9%, Synonym: ACN, Methyl cyanide. Acetonitrile (MeCN) may be used as an extraction solvent during the quantification of pesticide residues in fruits and vegetables.