Learn More
VWR INTERNATIONAL LLC Acetonitrile ≥99.5% for HPLC, 75-05-8, MFCD00001878, 4L
Catalog No. NC1313668
Encompass
Formula: H₃CCN, MW: 41.05 g/mol, Boiling Pt: 81.6 °C (1013 hPa), Melting Pt: -45.7 °C, Storage Temperature: Ambient, Synonyms: Methane, cyano- , Ethanoic acid nitrile , Methyl cyanide , Methanecarbonitrile , Ethanenitrile , Cyanomethane , CAN. High purity, functionally tested reagents suitable for HPLC and mass spectrometry. Low UV absorbing and low particulate count to reduce background.
Catalog No. NC1313668 Supplier VWR INTERNATIONAL LLC Supplier No. 97066096
Formula: H₃CCN, MW: 41.05 g/mol, Boiling Pt: 81.6 °C (1013 hPa), Melting Pt: -45.7 °C, Storage Temperature: Ambient, Synonyms: Methane, cyano- , Ethanoic acid nitrile , Methyl cyanide , Methanecarbonitrile , Ethanenitrile , Cyanomethane , CAN. High purity, functionally tested reagents suitable for HPLC and mass spectrometry. Low UV absorbing and low particulate count to reduce background.