Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile HPLC Plus 99.
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 349984X4L
Encompass
Acetonitrile (MeCN) is widely employed as a solvent in various studies and has high dielectric constant (37.5). Various purification procedures to obtain different grades of acetonitrile for use in different studies (polarography spectroscopy etc.) have been reported by many researchers.
Catalog No. 50-290-3243