Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019439 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9439 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9439 Supplier MilliporeSigma Supelco Supplier No. PHR15513X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Acetonitrile is an aliphatic nitrile, widely used as a solvent and intermediate in organic syntheses. It is also used in cosmetic products, and as a chemical intermediate in the manufacturing of pesticides and in industrial extraction process.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 75-05-8
Vapor Pressure 72.8 mmHg (20°C)
Vapor Density 1.41 (vs. air)
Density 0.786 g/mL (at 25°C)
Linear Formula CH3CN
MDL Number MFCD00001878
Melting Point -45°C
Boiling Point 81°C to 82°C
Synonym Acetonitrile; ACN; Cyanomethane; Ethyl nitrile; Methyl cyanide
Quantity 3 x 1.2 mL
Formula Weight 41.05
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.