Learn More
Andwin Scientific ACETONITRILE SFTY WSH BTL 6/PK
SDP
Catalog No. NC9571298
Encompass_Preferred
The Safety Wash Bottles are ideal for short-term dispensing purposes. This product features leakproof and break-resistant design. The bottles are sold with DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equ
Catalog No. NC9571298 Supplier Andwin Scientific Supplier No. 16649905
The Safety Wash Bottles are ideal for short-term dispensing purposes. This product features leakproof and break-resistant design. The bottles are sold with DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equ