Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile solution contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC | 75-05-8 | MFCD00001878 | 4X4L
Catalog No. 501858020
Encompass
Acetonitrile solution contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 4X4L
Catalog No. 50-185-8020 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6791864X4L
Acetonitrile solution contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 4X4L