Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile solution contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS) | 75-05-8 | MFCD00001878 | 4X4L
Catalog No. 501858029
Encompass
Acetonitrile solution contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS) | Purity: >=99.6% | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 4X4L
Catalog No. 50-185-8029 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9006864X4L
Acetonitrile solution contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS) | Purity: >=99.6% | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 4X4L