Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetonitrile solution contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC | 75-05-8 | MFCD00001878 | 20L
Catalog No. 501858014
Encompass
Acetonitrile solution contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 20L
Catalog No. 50-185-8014 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 57473220L
Acetonitrile solution contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC | Mol Wt: 41.05 | 75-05-8 | MFCD00001878 | 20L