Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Supelco

Catalog No. M1047262000 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1047262000 2 L
M1047260500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1047262000 Supplier MilliporeSigma Supelco Supplier No. 1047262000
Add to Cart
Add to Cart

Tested for PFAS Methods LiChrosolv™

  • Application Security: Quality control on 40 PFAS analytes according to EPA 533, EPA 537.1, and EPA 1633.
  • Time and Cost Savings: EPA 1633 requires batch validation unless the vendor tests for PFAS analytes listed in this method.
  • Lowest Impurity Profile: Ensures an interference-free baseline.
  • LC-MS Suitability: ESI/APCI (+) < 2 ppb; ESI/APCI (-) < 10 ppb.
  • Minimized Contamination: Borosilicate glass bottles reduce contamination with metal ions.
  • Trace Metal Impurity Control: Levels of trace metal impurities are minimized to <5 ppb, reducing metal ion adduct formation.
  • Microfiltration: Solvents are filtered through a 0.2 μm filter to ensure purity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 41.05

Specifications

CAS 75-05-8
Quantity 2 L
Boiling Point 81°C to 82°C (lit.)
Percent Purity ≥99.9% (GC)
Chemical Name or Material Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.