Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Biosciences Acetonitrile, Super Dehydrated, 3 L, H2O ≤10 ppm

Catalog No. NC3953033
Encompass_Preferred

FUJIFILM Wako Acetonitrile, Super Dehydrated is a high-purity, anhydrous organic solvent formulated for moisture-sensitive organic synthesis and analytical applications. The product is controlled to extremely low water content and is supplied in inert-gas-packed containers to preserve anhydrous quality.

  • Super-dehydrated grade with H2O ≤ 10 ppm.
  • Pack size: 3 L.
  • CAS number: 75-05-8.
  • Packed under inert gas and protected from light.
  • Suitable for organic synthesis and moisture-sensitive reactions.
  • Small-volume bottles feature a syringe-cap for withdrawal without opening.

Catalog No. NC3953033 Supplier FUJIFILM Biosciences Supplier No. 01422903
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FUJIFILM Wako Acetonitrile, Super Dehydrated is a high-purity, anhydrous organic solvent formulated for moisture-sensitive organic synthesis and analytical applications. The product is controlled to extremely low water content and is supplied in inert-gas-packed containers to preserve anhydrous quality.

  • Super-dehydrated grade with H2O ≤ 10 ppm.
  • Pack size: 3 L.
  • CAS number: 75-05-8.
  • Packed under inert gas and protected from light.
  • Suitable for organic synthesis and moisture-sensitive reactions.
  • Small-volume bottles feature a syringe-cap for withdrawal without opening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.