Learn More
Cayman Chemical Acetophenazine (maleate), Purity: ≥98%, CAS: 5714-00-1, 100 mg
SDP
Catalog No. 502946510
Encompass_Preferred
A typical antipsychotic; binds to 5-HT6 and 5-HT7 receptors and the androgen receptor (Kis = 72, 2.4, and 800 nM, respectively); inhibits the binding of SKF 10047 (Ki = 36 nM) and spiperone (IC50 = 17 nM) to guinea pig brain membranes; increases the duration of fighting and the latency to submission in mice
Catalog No. 50-294-6510 Supplier Cayman Chemical Supplier No. 34415100 MG
A typical antipsychotic; binds to 5-HT6 and 5-HT7 receptors and the androgen receptor (Kis = 72, 2.4, and 800 nM, respectively); inhibits the binding of SKF 10047 (Ki = 36 nM) and spiperone (IC50 = 17 nM) to guinea pig brain membranes; increases the duration of fighting and the latency to submission in mice