Learn More
Apexbio Technology LLC ACETOPHENONE-1G
SDP
Catalog No. 5000569773
Encompass_Preferred
Acetophenone (CAS No 98-86-2) is a versatile compound often employed in biomedical research particularly in pharmacological and toxicological studies Originating as a simple aromatic ketone it serves as a precursor in the metabolism of various pharmaceuticals Its action mechanism involves multiple targets including signaling pathways related to neural and enzymatic activities In vitro acetophenone demonstrates activity in the nanomolar to low micromolar range making it effective in modulating cellular responses It is frequently used in applications involving cell models like neuronal and liver cells as well as in animal studies for drug screening Experimental concentrations typically depend on the specific research design Acetophenone s ability to interact with diverse biological systems underlines its role as a critical tool in the study of drug actions and metabolic pathways
Catalog No. 50-005-69773 Supplier Apexbio Technology LLC Supplier No. C69691000
Acetophenone (CAS No 98-86-2) is a versatile compound often employed in biomedical research particularly in pharmacological and toxicological studies Originating as a simple aromatic ketone it serves as a precursor in the metabolism of various pharmaceuticals Its action mechanism involves multiple targets including signaling pathways related to neural and enzymatic activities In vitro acetophenone demonstrates activity in the nanomolar to low micromolar range making it effective in modulating cellular responses It is frequently used in applications involving cell models like neuronal and liver cells as well as in animal studies for drug screening Experimental concentrations typically depend on the specific research design Acetophenone s ability to interact with diverse biological systems underlines its role as a critical tool in the study of drug actions and metabolic pathways