Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acetophenone

Catalog No. A10701100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10701100ML 100 mL
A107011L 1 L
A107015ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A10701100ML Supplier Sigma Aldrich Supplier No. A10701100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetophenone

Specifications

CAS 98-86-2
Melting Point 19°C to 20°C
Density 1.03 g/mL at 25°C
Boiling Point 202°C
Molecular Formula C8H8O
Refractive Index n20/D 1.534
Linear Formula CH3COC6H5
MDL Number MFCD00008724
Quantity 100 mL
Synonym Methyl phenyl ketone
Molecular Weight (g/mol) 120.15 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.