Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020594 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0594 1 mL
11-101-9171 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0594 Supplier MilliporeSigma Supelco Supplier No. 421631MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetophenone is a colourless liquid with a sweet pungent taste. It can be used in perfume industry, as a fragrance ingredient and also as a flavouring agent in food industries.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetophenone
CAS 98-86-2
Ignition Point 1058°F
Vapor Pressure 1 mmHg (15°C)
Vapor Density 4.1 (vs. air)
Density 1.03 g/mL (at 25°C)
Flash Point 76°C
Physical Form Neat
Linear Formula CH3 COC6H5
Technique Gas Chromatography
MDL Number MFCD00008724
Melting Point 19°C to 20°C
Boiling Point 202°C
Synonym Methyl phenyl ketone
Type Analytical Standard
Industry Type Laboratory Use
Molecular Weight (g/mol) 120.15
Quantity 1 mL
Formula Weight 120.15
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.