Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone (Certified ACS), Fisher Chemical

Catalog No. A22500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A22-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A22-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 98-86-2
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

Melting Point 19.6°C
Color Yellow
Density 1.028g/cm³
Boiling Point 202°C
Flash Point 77°C
Packaging Glass Bottle
Linear Formula CH3COC6H5
Quantity 500 mL
Specific Gravity 1.023 to 1.027 (at 25°C)
Vapor Pressure 1.3mbar at 20°C
Formula Weight 120.15
Grade Certified
Physical Form Liquid
Chemical Name or Material Acetophenone
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Irritating to eyes. May cause skin and respiratory tract irritation. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Do not breathe vapors or spray mist. Avoid contact with skin, eyes and clothing. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:2
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.