Learn More
Medchemexpress LLC HY-W009884 100mg Medchemexpress, Acetosyringone CAS:2478-38-8 Purity:>98%
SDP
Catalog No. 501934529
Encompass_Preferred
Medchemexpress, HY-W009884 100mg Acetosyringone CAS:2478-38-8 Acetosyringone is a phenolic compound from wounded plant cells, enables virA gene which encodes a membrane-bound kinase to phosphorylate itself and activate the virG gene product, which stimulates the transcription of other vir genes and itself [1] . Acetosyringone enhances efficient Dunaliella transformation of Agrobacterium strains [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4529 Supplier Medchemexpress LLC Supplier No. HYW009884100MG
Medchemexpress, HY-W009884 100mg Acetosyringone CAS:2478-38-8 Acetosyringone is a phenolic compound from wounded plant cells, enables virA gene which encodes a membrane-bound kinase to phosphorylate itself and activate the virG gene product, which stimulates the transcription of other vir genes and itself [1] . Acetosyringone enhances efficient Dunaliella transformation of Agrobacterium strains [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.