Learn More
Apexbio Technology LLC Acetosyringone 2478-38-8 200mg
SDP
Catalog No. 5000092447
Encompass_Preferred
Acetosyringone is a phenolic small molecule that interacts with the Agrobacterium tumefaciens virulence (vir) regulatory system It functions by activating the membrane-bound kinase encoded by the virA gene initiating autophosphorylation events and subsequent activation of the virG transcriptional regulator which induces expression of additional virulence genes Acetosyringone exerts its biological activity primarily through the activation of the VirA/VirG two-component system Based on these properties acetosyringone is widely used in plant biotechnology for enhancing Agrobacterium-mediated transformation and genetic engineering studies in various plant species and algae
Catalog No. 50-000-92447 Supplier Apexbio Technology LLC Supplier No. C6580200
Acetosyringone is a phenolic small molecule that interacts with the Agrobacterium tumefaciens virulence (vir) regulatory system It functions by activating the membrane-bound kinase encoded by the virA gene initiating autophosphorylation events and subsequent activation of the virG transcriptional regulator which induces expression of additional virulence genes Acetosyringone exerts its biological activity primarily through the activation of the VirA/VirG two-component system Based on these properties acetosyringone is widely used in plant biotechnology for enhancing Agrobacterium-mediated transformation and genetic engineering studies in various plant species and algae