Learn More
Bioworld Acetosyringone , 1 g
Catalog No. 50255291
Encompass_Preferred
Itis a volatile chemical compound related to acetophenone and is a natural product that can be found in Posidonia oceanica.The compound is also produced by the male leaffooted bug and used in its communication system. This substance is involved in plant-pathogen recognition. In vitro studies show that acetosyringone increases mycorrhizae formation in the fungus Glomus intraradices. Total synthesis of acetosyringone has been performed by Crawford et al in 1956.Specifications:Synonym: 4'-Hydoxy-3'5'-dimethoxyacetophenoneIUPAC_Name: 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanoneRTECS Number: AM8440000
Catalog No. 50-255-291 Supplier Bioworld Supplier No. 401002971
Itis a volatile chemical compound related to acetophenone and is a natural product that can be found in Posidonia oceanica.The compound is also produced by the male leaffooted bug and used in its communication system. This substance is involved in plant-pathogen recognition. In vitro studies show that acetosyringone increases mycorrhizae formation in the fungus Glomus intraradices. Total synthesis of acetosyringone has been performed by Crawford et al in 1956.Specifications:Synonym: 4'-Hydoxy-3'5'-dimethoxyacetophenoneIUPAC_Name: 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanoneRTECS Number: AM8440000