Learn More
Cayman Chemical Acetosyringone, Purity: ≥98%, CAS: 2478-38-8, 1 g
SDP
Catalog No. 502993839
Encompass_Preferred
A phenol secreted by wounded plant tissues; induces expression of virulence A genes and chemotaxis in A. tumefaciens strains that contain a tumor-inducing plasmid used to transfer genetic information to plant cells; widely used to increase efficacy of genetic transformation for the creation of genetically modified dicotyledonous and monocotyledonous plants
Catalog No. 50-299-3839 Supplier Cayman Chemical Supplier No. 232241 G
A phenol secreted by wounded plant tissues; induces expression of virulence A genes and chemotaxis in A. tumefaciens strains that contain a tumor-inducing plasmid used to transfer genetic information to plant cells; widely used to increase efficacy of genetic transformation for the creation of genetically modified dicotyledonous and monocotyledonous plants