Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetovanillone 98 FG10KG
Catalog No. 502908308
Encompass
Acetovanillone is a structural analog of vanillin that occurs mainly in wine aged in oak barrels. It is a potent NADPH oxidase inhibitor.
Catalog No. 50-290-8308 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W50845410KG
Acetovanillone is a structural analog of vanillin that occurs mainly in wine aged in oak barrels. It is a potent NADPH oxidase inhibitor.