Learn More
Medchemexpress LLC Acetoxymethyl 2-(5-phenyl-1H-indol-3-yl)acetate | 2990123-10-7 | 99.0% | C19H17NO4 | 10MG
SDP
Catalog No. 5000259069
Encompass_Preferred
5-Ph-IAA-AM is an eggshell-permeable acetoxymethyl derivative of 5-Ph-IAA designed for use in auxin-inducible degron (AID2) systems. It provides enhanced protein degradation in laid C. elegans embryos, enabling temporally controlled depletion of target proteins to study embryogenesis and developmental processes.
  • Eggshell-permeant analog facilitates delivery into laid embryos.
  • Enhances protein degradation efficiency in AID2 systems.
  • Compatible with OsTIR1(F74G)-mediated auxin-inducible degron assays.
  • High purity supports reproducible experimental results.
  • Powder is stable under recommended cold storage conditions.
Catalog No. 50-002-59069 Supplier Medchemexpress LLC Supplier No. HY14189410MG
5-Ph-IAA-AM is an eggshell-permeable acetoxymethyl derivative of 5-Ph-IAA designed for use in auxin-inducible degron (AID2) systems. It provides enhanced protein degradation in laid C. elegans embryos, enabling temporally controlled depletion of target proteins to study embryogenesis and developmental processes.
  • Eggshell-permeant analog facilitates delivery into laid embryos.
  • Enhances protein degradation efficiency in AID2 systems.
  • Compatible with OsTIR1(F74G)-mediated auxin-inducible degron assays.
  • High purity supports reproducible experimental results.
  • Powder is stable under recommended cold storage conditions.