Learn More
Cayman Chemical AcetoxyvalerenIc AcId 10mg
SDP
Catalog No. 502991112
Encompass_Preferred
A sesquiterpenic acid; cytotoxic to GLC-4 and COLO 320 cells (IC50s = 111 and 124 µM, respectively)
Catalog No. 50-299-1112 Supplier Cayman Chemical Supplier No. 3151610 MG
A sesquiterpenic acid; cytotoxic to GLC-4 and COLO 320 cells (IC50s = 111 and 124 µM, respectively)