Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eMolecules​ TRC Acetyl-1,2-13C2 Coenzyme A Sodium Salt (~2% Unlabelled), Neat, Carbon

Supplier:  eMolecules​ 866849471

Encompass_Preferred

This product is Acetyl-1,2-13C2 Coenzyme A Sodium Salt (~2% Unlabelled), a stable isotope labelled analytical standard. It is provided in a neat format and is suitable for various laboratory applications.

  • Molecular formula: C2113C2H38N7O17P3S · x(Na)
  • Molecular weight: 811.562299
  • Storage temperature: −20°C
  • Shipping temperature: Room temperature
  • Country of origin: Canada
  • Product format: Neat
  • SIL type: Carbon
  • Product type: Impurity, stable isotope labelled, bioactive small molecule

Catalog No. NC3974590


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.