Learn More
Cell Signaling Technology Acetyl-Alpha-Tubulin Matched Antibody Pair, Human, Mouse, Rat, 1 Kit
SDP
Catalog No. 503132446
Encompass
The Acetyl-α-Tubulin (Lys40) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of mouse monoclonal alpha-Tubulin (DM1A) antibody and rabbit monoclonal Acetyl-alpha-Tubulin (Lys40) (D20G3) antibody.
  • Supplied in BSA and azide-free 1X PBS formulation.
  • Validated for use in human, mouse, and rat species.
  • Optimized for applications including ELISA.
  • Requires user-determined optimal dilutions and concentrations.
Catalog No. 50-313-2446 Supplier Cell Signaling Technology Supplier No. 53362S
The Acetyl-α-Tubulin (Lys40) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of mouse monoclonal alpha-Tubulin (DM1A) antibody and rabbit monoclonal Acetyl-alpha-Tubulin (Lys40) (D20G3) antibody.
  • Supplied in BSA and azide-free 1X PBS formulation.
  • Validated for use in human, mouse, and rat species.
  • Optimized for applications including ELISA.
  • Requires user-determined optimal dilutions and concentrations.