Learn More
CPC Scientific Ac-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr-Lys-Lys-Gly-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ENDP002A
Encompass
ONE-LETTER SEQUENCE: Ac-YGGFMTSEKSQTPLVTLFKNAIIKNAYKKGEMOLECULAR FORMULA: C160H253N39O47S1MOLECULAR WEIGHT:3507.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Pain
Catalog No. 50-210-9226