Learn More
Bachem Acetyl-Calpastatin (184-210) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4027881.1000
Encompass_Preferred
Acetyl-Calpastatin (184-210) (human) Trifluoroacetate, Ac-Asp-Pro-Met-Ser-Ser-Thr-Tyr-Ile-Glu-Glu-Leu-Gly-Lys-Arg-Glu-Val-Thr-Ile-Pro-Pro-Lys-Tyr-Arg-Glu-Leu-Leu-Ala-NH2, 123714-50-1, C142H230N36O44S, Mr 3177.67, 1mg. Acetyl-Calpain Inhibitor Fragment (184-210) (human), Acetyl-Sperm BS-17 Component (184-210) (human). Acetyl-calpastatin 27 peptide, AC27P, a calpain inhibitor homologous to the minimal inhibitory domain I of human calpastatin, was shown to modulate the amyloid b-protein 42 secretion ratio. This finding might point to the possibility of exploring compounds that inhibit Ab42 secretion.
Catalog No. 50-194-6642