Learn More
Apexbio Technology LLC ACETYL-CARNITINE-D3-HCL-5MG
SDP
Catalog No. 5000570687
Encompass_Preferred
Acetyl carnitine-d3 HCl (CAS No 1334532-17-0) is a deuterated form of acetyl carnitine used in biomedical research to study cellular metabolism and energy production It modulates various biochemical pathways by facilitating the transport of fatty acids into mitochondria enhancing fatty acid oxidation and increasing cellular energy availability Its activity is observed in nanomolar to low micromolar ranges Typical applications involve cell models and animal studies particularly for research in metabolic disorders and mitochondrial functions Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70687 Supplier Apexbio Technology LLC Supplier No. SL10125
Acetyl carnitine-d3 HCl (CAS No 1334532-17-0) is a deuterated form of acetyl carnitine used in biomedical research to study cellular metabolism and energy production It modulates various biochemical pathways by facilitating the transport of fatty acids into mitochondria enhancing fatty acid oxidation and increasing cellular energy availability Its activity is observed in nanomolar to low micromolar ranges Typical applications involve cell models and animal studies particularly for research in metabolic disorders and mitochondrial functions Experimental concentrations typically depend on the specific research design