Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acetyl Chloride Solution

Catalog No. 708496100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-849-6100ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-849-6100ML Supplier Sigma Aldrich Supplier No. 708496100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetyl chloride (AcCl) can be used as: An acylating reagent to prepare useful key intermediates in organic synthesis. A reagent to convert oxathioacetals and dithioacetals into corresponding carbonyl compounds in the presence of sodium nitrite. An activating or acylating reagent in solid-phase organic synthesis. A reagent to prepare various alkyl and aryl tetrahydropyranyl ethers from corresponding alcohols.

Specifications

CAS 925706-40-7
Melting Point 200°C to 206°C
Molecular Formula C23H28BF4N3O
Linear Formula C23H28BF4N3O
Refractive Index n20/D 1.423
MDL Number MFCD16621403
Quantity 100 mg
Molecular Weight (g/mol) 449.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.