Learn More
Cusabio Technology LLC Acetyl-CoA carboxylase 1
Catalog No. 5000048227
Encompass_Preferred
This kit detects ACACA (Acetyl-CoA carboxylase 1) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA methodology It offers a quantitative detection range of 9 38 pg/mL to 600 pg/mL with a sensitivity of 2 34 pg/mL ensuring precise measurement of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Results are determined by absorbance measurement at 450 nm providing a reliable and standardized approach for ACACA analysis in research or diagnostic applications The kit s optimized protocol maintains specificity and reproducibility across diverse biological matrices making it suitable for studies involving lipid metabolism regulation enzyme activity monitoring or biomarker discovery related to ACACA expression
Catalog No. 50-000-48227 Supplier Cusabio Technology LLC Supplier No. CSBEL001119HU
This kit detects ACACA (Acetyl-CoA carboxylase 1) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA methodology It offers a quantitative detection range of 9 38 pg/mL to 600 pg/mL with a sensitivity of 2 34 pg/mL ensuring precise measurement of low-abundance samples The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 L per test Results are determined by absorbance measurement at 450 nm providing a reliable and standardized approach for ACACA analysis in research or diagnostic applications The kit s optimized protocol maintains specificity and reproducibility across diverse biological matrices making it suitable for studies involving lipid metabolism regulation enzyme activity monitoring or biomarker discovery related to ACACA expression