Learn More
Medchemexpress LLC Acetyl CoA Carboxylase 1 (ACC1) Antibody (YA646) | 266 kDa | 50 UL
SDP
Catalog No. 5000395876
Encompass_Preferred
This antibody is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Acetyl CoA Carboxylase 1 (ACC1). It is suitable for applications such as Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P).
  • Derived from rabbit host.
  • Reacts with human and mouse samples.
  • Monoclonal antibody for consistent results.
  • Predicted molecular weight of 266 kDa.
  • Supplied at a concentration of 1 mg/mL.
  • Formulated in 1*TBS (pH 7.4), 0.05% BSA, and 40% Glycerol, including 0.05% Sodium Azide as a preservative.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Shipped with blue ice to maintain stability.
Catalog No. 50-003-95876 Supplier Medchemexpress LLC Supplier No. HYP8048250UL
This antibody is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Acetyl CoA Carboxylase 1 (ACC1). It is suitable for applications such as Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P).
  • Derived from rabbit host.
  • Reacts with human and mouse samples.
  • Monoclonal antibody for consistent results.
  • Predicted molecular weight of 266 kDa.
  • Supplied at a concentration of 1 mg/mL.
  • Formulated in 1*TBS (pH 7.4), 0.05% BSA, and 40% Glycerol, including 0.05% Sodium Azide as a preservative.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Shipped with blue ice to maintain stability.