Learn More
Medchemexpress LLC Acetyl-CoA Carboxylase-IN-1 | 179343-23-8 | C13H9Br2N5 | 1 ML
SDP
Catalog No. 5000374163
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM. It also possesses antibacterial activity, making it suitable for studies on bacterial infections.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor
  • IC50 value of <5 nM
  • Possesses antibacterial activity
  • Effective against various E. coli strains
Catalog No. 50-003-74163 Supplier Medchemexpress LLC Supplier No. HY15211710MM/1ML
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM. It also possesses antibacterial activity, making it suitable for studies on bacterial infections.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor
  • IC50 value of <5 nM
  • Possesses antibacterial activity
  • Effective against various E. coli strains