Learn More
Medchemexpress LLC Acetyl-CoA Carboxylase-IN-1 | 179343-23-8 | C13H9Br2N5 | 100 MG
SDP
Catalog No. 5000374160
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM. It also exhibits antibacterial activity. This product is for research use only and not intended for sale to patients.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM
  • Exhibits antibacterial activity
  • Demonstrates antibacterial activity against E. coli with varying MIC values depending on the strain
Catalog No. 50-003-74160 Supplier Medchemexpress LLC Supplier No. HY152117100MG
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM. It also exhibits antibacterial activity. This product is for research use only and not intended for sale to patients.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM
  • Exhibits antibacterial activity
  • Demonstrates antibacterial activity against E. coli with varying MIC values depending on the strain