Learn More
Medchemexpress LLC Acetyl-CoA Carboxylase-IN-1 | 179343-23-8 | C13H9Br2N5 | 25 MG
SDP
Catalog No. 5000374379
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM, demonstrated to exhibit antibacterial activity. It is available as an off-white to light yellow solid powder and is soluble in DMSO, making it suitable for various laboratory applications.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM
  • Exhibits antibacterial activity against E. coli
  • Soluble in DMSO for in vitro applications
  • Long-term stability when stored as a powder at -20°C (3 years) or 4°C (2 years)
  • Stable in solvent at -80°C (6 months) or -20°C (1 month)
Catalog No. 50-003-74379 Supplier Medchemexpress LLC Supplier No. 25MGHY152117
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM, demonstrated to exhibit antibacterial activity. It is available as an off-white to light yellow solid powder and is soluble in DMSO, making it suitable for various laboratory applications.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of less than 5 nM
  • Exhibits antibacterial activity against E. coli
  • Soluble in DMSO for in vitro applications
  • Long-term stability when stored as a powder at -20°C (3 years) or 4°C (2 years)
  • Stable in solvent at -80°C (6 months) or -20°C (1 month)