Learn More
Medchemexpress LLC Acetyl-CoA Carboxylase-IN-1 | 179343-23-8 | C13H9Br2N5 | 5 MG
SDP
Catalog No. 5000373472
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM. It also demonstrates antibacterial activity.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM.
  • Exhibits antibacterial activity.
  • Purity: 99.18%.
Catalog No. 50-003-73472 Supplier Medchemexpress LLC Supplier No. HY1521175MG
Acetyl-CoA Carboxylase-IN-1 is a potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM. It also demonstrates antibacterial activity.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor with an IC50 value of <5 nM.
  • Exhibits antibacterial activity.
  • Purity: 99.18%.