Learn More
Medchemexpress LLC Acetyl-CoA Carboxylase-IN-1 | 179343-23-8 | C13H9Br2N5 | 50 MG
SDP
Catalog No. 5000373648
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent inhibitor of acetyl-CoA carboxylase (ACC), demonstrating an IC50 value of less than 5 nM. This compound also exhibits significant antibacterial activity, with in vitro studies on *E. coli* showing minimum inhibitory concentration (MIC) values ranging from 0.1 μM to over 25 μM depending on the bacterial strain.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor
  • Exhibits antibacterial activity against *E. coli*
  • High purity of 99.18%
  • Available as an off-white to light yellow solid
  • Long-term stability as a powder at -20°C for up to 3 years
  • Stable in solvent at -80°C for 6 months
Catalog No. 50-003-73648 Supplier Medchemexpress LLC Supplier No. HY15211750MG
Acetyl-CoA Carboxylase-IN-1 is a potent inhibitor of acetyl-CoA carboxylase (ACC), demonstrating an IC50 value of less than 5 nM. This compound also exhibits significant antibacterial activity, with in vitro studies on *E. coli* showing minimum inhibitory concentration (MIC) values ranging from 0.1 μM to over 25 μM depending on the bacterial strain.
  • Potent acetyl-CoA carboxylase (ACC) inhibitor
  • Exhibits antibacterial activity against *E. coli*
  • High purity of 99.18%
  • Available as an off-white to light yellow solid
  • Long-term stability as a powder at -20°C for up to 3 years
  • Stable in solvent at -80°C for 6 months