Learn More
Medchemexpress LLC Acetyl-CoA carboxylase-IN-1 | 179343-23-8 | MFCD12828288 | 99.2% | 395.05 g·mol⁻¹ | C13H9Br2N5 | 10 MG
SDP
Catalog No. 5000287680
Encompass_Preferred
Acetyl-CoA Carboxylase-IN-1 is a potent small-molecule inhibitor of acetyl-CoA carboxylase used for biochemical and microbiology research. It displays potent ACC inhibition (IC50 ≈ 5 nM) and reported antibacterial activity against Escherichia coli under defined in vitro assay conditions. The material is supplied as a solid (off-white to light yellow) for research use only and should be stored and handled according to the supplier's SDS.
  • Potent ACC inhibitor with IC50 ≈ 5 nM.
  • Antibacterial activity reported against Escherichia coli in vitro.
  • Chemical identifiers: CAS 179343-23-8; formula C13H9Br2N5; MW 395.05 g·mol⁻¹.
  • High purity (manufacturer-reported ~99.18%).
  • Available in small research pack sizes (e.g., 10 mg) as a solid.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
Catalog No. 50-002-87680 Supplier Medchemexpress LLC Supplier No. HY15211710MG
Acetyl-CoA Carboxylase-IN-1 is a potent small-molecule inhibitor of acetyl-CoA carboxylase used for biochemical and microbiology research. It displays potent ACC inhibition (IC50 ≈ 5 nM) and reported antibacterial activity against Escherichia coli under defined in vitro assay conditions. The material is supplied as a solid (off-white to light yellow) for research use only and should be stored and handled according to the supplier's SDS.
  • Potent ACC inhibitor with IC50 ≈ 5 nM.
  • Antibacterial activity reported against Escherichia coli in vitro.
  • Chemical identifiers: CAS 179343-23-8; formula C13H9Br2N5; MW 395.05 g·mol⁻¹.
  • High purity (manufacturer-reported ~99.18%).
  • Available in small research pack sizes (e.g., 10 mg) as a solid.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).