Learn More
Medchemexpress LLC Acetyl-CoA synthetas 50ug
SDP
Catalog No. 5000196808
Encompass_Preferred
Recombinant mouse acetyl-CoA synthetase 2 (ACSS2), expressed in E. coli and purified with an N-terminal His tag. Supplied in microgram quantities for biochemical and enzymatic studies, the enzyme converts acetate to acetyl-CoA and is provided with purity, molecular weight, and storage information for reliable experimental use.
  • Purified recombinant protein with >90% purity by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Approximate molecular weight of 82.9 kDa for assay planning.
  • Available in microgram sizes suitable for biochemical assays.
  • Expressed in E. coli and supplied with COA and SDS documentation.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term storage.
Catalog No. 50-001-96808 Supplier Medchemexpress LLC Supplier No. HYP7164250UG
Recombinant mouse acetyl-CoA synthetase 2 (ACSS2), expressed in E. coli and purified with an N-terminal His tag. Supplied in microgram quantities for biochemical and enzymatic studies, the enzyme converts acetate to acetyl-CoA and is provided with purity, molecular weight, and storage information for reliable experimental use.
  • Purified recombinant protein with >90% purity by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Approximate molecular weight of 82.9 kDa for assay planning.
  • Available in microgram sizes suitable for biochemical assays.
  • Expressed in E. coli and supplied with COA and SDS documentation.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term storage.