Learn More
Medchemexpress LLC Acetyl-CoA synthetas 5ug
SDP
Catalog No. 5000196064
Encompass_Preferred
Recombinant mouse Acetyl-CoA synthetase 1 (ACSS2) is supplied as an N-terminal 6xHis-tagged protein produced in E. coli for biochemical and metabolic research. The protein is provided in microgram quantities with purity >90% (example COA 92%) and an approximate molecular weight of 82.9 kDa, and is intended for use in enzymatic assays, binding studies, and as a reference standard.
  • His-tagged recombinant protein for easy purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Approximately 82.9 kDa molecular weight for accurate size reference.
  • Produced in E. coli for efficient expression.
  • Suitable for enzymatic assays and metabolic studies.
  • Provided in small microgram quantities for research use.
  • Certificate of analysis available for batch verification.
Catalog No. 50-001-96064 Supplier Medchemexpress LLC Supplier No. HYP716425UG
Recombinant mouse Acetyl-CoA synthetase 1 (ACSS2) is supplied as an N-terminal 6xHis-tagged protein produced in E. coli for biochemical and metabolic research. The protein is provided in microgram quantities with purity >90% (example COA 92%) and an approximate molecular weight of 82.9 kDa, and is intended for use in enzymatic assays, binding studies, and as a reference standard.
  • His-tagged recombinant protein for easy purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Approximately 82.9 kDa molecular weight for accurate size reference.
  • Produced in E. coli for efficient expression.
  • Suitable for enzymatic assays and metabolic studies.
  • Provided in small microgram quantities for research use.
  • Certificate of analysis available for batch verification.