Learn More
Medchemexpress LLC Acetyl-CoA synthetase | 9012-31-1 | 5 MG
SDP
Catalog No. 5000399492
Encompass_Preferred
Acetyl-CoA synthetase (ACS) is a key enzyme that catalyzes the conversion of acetate to acetyl-CoA (Ac-CoA). It catalyzes the formation of thioester bonds between coenzyme A and carboxylic acids, while simultaneously hydrolyzing ATP into AMP and pyrophosphate.
  • Key enzyme for acetate to acetyl-CoA conversion
  • Catalyzes thioester bond formation and ATP hydrolysis
  • Appears as a colorless to off-white solid-liquid mixture
  • Classified as an enzyme, involved in chemical synthesis
  • Relevant to cancer metabolism and related research
Catalog No. 50-003-99492 Supplier Medchemexpress LLC Supplier No. HYP28275MG
Acetyl-CoA synthetase (ACS) is a key enzyme that catalyzes the conversion of acetate to acetyl-CoA (Ac-CoA). It catalyzes the formation of thioester bonds between coenzyme A and carboxylic acids, while simultaneously hydrolyzing ATP into AMP and pyrophosphate.
  • Key enzyme for acetate to acetyl-CoA conversion
  • Catalyzes thioester bond formation and ATP hydrolysis
  • Appears as a colorless to off-white solid-liquid mixture
  • Classified as an enzyme, involved in chemical synthesis
  • Relevant to cancer metabolism and related research