Learn More
Cayman Chemical Acetyl Coenzyme A (sodium salt), 102029-73-2, 25 mg
SDP
Supplier:  Cayman Chemical 1616025
Encompass_Preferred
A pivotal molecule in the Krebs cycle, in the synthesis of fatty acids, and in isoprenoid-based protein modifications; serves as an intermediate in oxidation of fatty acids and amino acids; an essential cofactor or substrate for acetyltransferases and acyltransferases
Catalog No. NC1247664