Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetyl coenzyme A sodium salt, 102029-73-2, MFCD00078858, 1 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A20561MG
Encompass
Empirical Formula (Hill Notation): C23H38N7O17P3S, Molecular Weight: 809.57, ≥93% (HPLC), powder, Synonym: Acetyl CoA, Acetyl-S- CoA. An essential cofactor in enzymatic acetyl transfer reactions.
Catalog No. NC0723795