Learn More
Cayman Chemical Acetyl-Coenzyme A-d3 (sodium salt), Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502943304
Encompass_Preferred
An internal standard for the quantification of acetyl-coenzyme A by GC- or LC-MS
Catalog No. 50-294-3304 Supplier Cayman Chemical Supplier No. 404581 MG
An internal standard for the quantification of acetyl-coenzyme A by GC- or LC-MS