Learn More
Medchemexpress LLC HY-113596 10mg , Acetyl Coenzyme A (trisodium) CAS:102029-73-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11359610MG
Encompass_Preferred
HY-113596 10mg Acetyl Coenzyme A (trisodium) CAS: 102029-73-2 Acetyl Coenzyme A trisodium (Acetyl-CoA trisodium) is a central metabolic intermediate. Acetyl Coenzyme A trisodium is the actual molecule through which glycolytic pyruvate enters the tricarboxylic acid (TCA) cycle, is a key precursor of lipid synthesis, and is the sole donor of the acetyl groups for acetylation. Acetyl Coenzyme A trisodium acts as a potent allosteric activator of pyruvate carboxylase (PC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0560