Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetyl-(D-Phe2,Lys15,Arg16,Leu27)-VIP (1-7)-GRF (8-27) | 202463-00-1 | 98.0% | 3273.83 | 1 MG
SDP

Catalog No. 5000398498
Encompass_Preferred

Acetyl-(D-Phe2,Lys15,Arg16,Leu27)-VIP (1-7)-GRF (8-27) is a vasoactive intestinal peptide (VIP) and a VIP1 antagonist.

  • Formula: C150H246N44O38
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ac-His-{d-Phe}-Asp-Ala-Val-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Lys-Arg-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Leu-NH2
  • Shortened sequence: Ac-H-{d-Phe}-DAVFTNSYRKVLKRLSARKLLQDIL-NH2
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Sealed storage, away from moisture; powder: -80°C for 2 years, -20°C for 1 year; in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture).
  • Solubility: H2O: 33.33 mg/mL (10.18 mM; adjust pH to 2 with 1 M HCl).

Catalog No. 50-003-98498 Supplier Medchemexpress LLC Supplier No. HYP47761MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetyl-(D-Phe2,Lys15,Arg16,Leu27)-VIP (1-7)-GRF (8-27) is a vasoactive intestinal peptide (VIP) and a VIP1 antagonist.

  • Formula: C150H246N44O38
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ac-His-{d-Phe}-Asp-Ala-Val-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Lys-Arg-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Leu-NH2
  • Shortened sequence: Ac-H-{d-Phe}-DAVFTNSYRKVLKRLSARKLLQDIL-NH2
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Sealed storage, away from moisture; powder: -80°C for 2 years, -20°C for 1 year; in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture).
  • Solubility: H2O: 33.33 mg/mL (10.18 mM; adjust pH to 2 with 1 M HCl).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.