Learn More
Abcam N-Acetyl-D-glucosaminitol, 1G
Supplier:  Abcam AB2750521G
Encompass_Preferred
MW 223.22 Da. High purity reduced analog of N-Acetyl-D-Glucosamine, useful in bacterial selective media. Differentiates bacteria based on metabolic characteristics. Standard for certain carbohydrate detection systems.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-211