Learn More
Epigentek Inc Histone H3K56ac (Acetyl H3K56) Polyclonal Antibody (50 ul)
SDP
Supplier:  Epigentek Inc A4026050
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human Acetyl-Histone H3-K56. Tested Applications: WB, IHC, IF, IP, ChIP, ChIPseq. Reactivity: Mouse, Rat, Human, Broad Range. Alternative Names: H3K56ac antibody, H3K56a antibody.
Catalog No. 50-109-5495