Learn More
Cell Signaling Technology Acetyl-Histone Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729487
Encompass
The Acetyl-Histone Antibody Sampler Kit provides a fast and economical solution for evaluating the acetylation states of histones H2A, H2B, H3, and H4. This kit contains primary and secondary antibodies sufficient for performing two Western blot experiments.
  • Includes primary antibodies for acetylated histones and total histone controls
  • Contains HRP-linked secondary antibody for detection
  • Supplied in storage buffer with 50% glycerol
  • Validated for multiple applications including Western blotting, immunofluorescence, and chromatin immunoprecipitation
Catalog No. 50-272-9487 Supplier Cell Signaling Technology Supplier No. 9933T
The Acetyl-Histone Antibody Sampler Kit provides a fast and economical solution for evaluating the acetylation states of histones H2A, H2B, H3, and H4. This kit contains primary and secondary antibodies sufficient for performing two Western blot experiments.
  • Includes primary antibodies for acetylated histones and total histone controls
  • Contains HRP-linked secondary antibody for detection
  • Supplied in storage buffer with 50% glycerol
  • Validated for multiple applications including Western blotting, immunofluorescence, and chromatin immunoprecipitation