Learn More
Cell Signaling Technology Acetyl-Histone H2B Matched Antibody Pair, ELISA, Unconjugated, 1 Kit
SDP
Catalog No. 503134062
Encompass
The Acetyl-Histone H2B Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This pair is compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads for use on platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8
  • Formulated without BSA or azide
  • Validated for reactivity in human, mouse, and monkey samples
  • Storage recommended at -20°C in single-use aliquots to minimize freeze/thaw cycles
Catalog No. 50-313-4062 Supplier Cell Signaling Technology Supplier No. 44879S
The Acetyl-Histone H2B Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This pair is compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads for use on platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8
  • Formulated without BSA or azide
  • Validated for reactivity in human, mouse, and monkey samples
  • Storage recommended at -20°C in single-use aliquots to minimize freeze/thaw cycles