Learn More
ABclonal Technology Acetyl-Histone H3-K27
SDP
Supplier:  ABclonal Technology A2771100UL
Encompass_Preferred
Core component of nucleosome Nucleosomes wrap and compact DNA into chromatin limiting DNA accessibility to the cellular machineries which require DNA as a template Histones thereby play a central role in transcription regulation DNA repair DNA replication and chromosomal stability DNA accessibility is regulated via a complex set of post-translational modifications of histones also called histone code and nucleosome remodeling
Catalog No. 50-000-14968