Learn More
ABclonal Technology Acetyl-Histone H3-K9
SDP
Catalog No. 5000008004
Encompass_Preferred
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes This structure consists of approximately 146 bp of DNA wrapped around a nucleosome an octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is found in the large histone gene cluster on chromosome 6p22-p21 3
Catalog No. 50-000-08004 Supplier ABclonal Technology Supplier No. A21107100UL
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes This structure consists of approximately 146 bp of DNA wrapped around a nucleosome an octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is found in the large histone gene cluster on chromosome 6p22-p21 3